
Ioxynil-methyl

Synonym(s):
3,5-Diiodo-4-methoxybenzonitrile
Empirical Formula (Hill Notation):
C8H5I2NO
CAS Number:
Molecular Weight:
384.94
Products
Shipping
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing