Skip to Content
Merck
Imiquimod Related Compound B pharmaceutical secondary standard, certified reference material

Imiquimod Related Compound B

Imiquimod Related Compound B pharmaceutical secondary standard, certified reference material
Synonym(s):
1-Isobutyl-1H-imidazo[4,5-c]quinoline 5-oxide
Empirical Formula (Hill Notation):
C14H15N3O
CAS Number:
Molecular Weight:
241.29

Products

Shipping
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade