![Imiquimod Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/123/369/8806043b-8760-4922-bc29-fc5a970bc19c/640/8806043b-8760-4922-bc29-fc5a970bc19c.jpg)
Imiquimod Related Compound A
![Imiquimod Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/123/369/8806043b-8760-4922-bc29-fc5a970bc19c/640/8806043b-8760-4922-bc29-fc5a970bc19c.jpg)
Synonym(s):
1-Isobutyl-1H-imidazo[4,5-c]quinoline
Empirical Formula (Hill Notation):
C14H15N3
CAS Number:
Molecular Weight:
225.29
Products
Shipping
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2