Skip to Content
Merck
Hydroquinine-9-phenanthryl ether 97%

Hydroquinine-9-phenanthryl ether

Hydroquinine-9-phenanthryl ether 97%
Synonym(s):
O-(9-Phenanthryl)hydroquinine
Empirical Formula (Hill Notation):
C34H34N2O2
CAS Number:
Molecular Weight:
502.65

Products

Shipping
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing