Hydrocortisone hemisuccinate
Synonym(s):
Hydrocortisone 21-hemisuccinate monohydrate, 11β,17aα,21-Trihydroxy-4-pregnene-3,20-dione 21-hemisuccinate
Empirical Formula (Hill Notation):
C25H34O8 · H2O
CAS Number:
Molecular Weight:
480.55
Products
Shipping
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2