Homopiperazine-1,4-bis(2-ethanesulfonic acid)
Synonym(s):
Hexahydro-1H-1,4-diazepine-1,4-bis(2-ethanesulfonic acid), Homo-PIPES
Empirical Formula (Hill Notation):
C9H20N2O6S2
CAS Number:
Molecular Weight:
316.39
Products
Shipping
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing