Skip to Content
Merck
Hexahydro-4-methylphthalic anhydride, mixture of cis and trans 96%

Hexahydro-4-methylphthalic anhydride, mixture of cis and trans

Hexahydro-4-methylphthalic anhydride, mixture of cis and trans 96%
Synonym(s):
4-Methyl-1,2-cyclohexanedicarboxylic anhydride
Empirical Formula (Hill Notation):
C9H12O3
CAS Number:
Molecular Weight:
168.19
EC Number:

Products

Shipping
Product Category
Formula Weight
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing