Hexadecafluorodecanedioic acid
Synonym(s):
Hexadecafluorosebacic acid, Perfluorodecanedioic acid, Perfluorosebacic acid
Linear Formula:
HOOC(CF2)8COOH
CAS Number:
Molecular Weight:
490.09
Products
Shipping
Product Category
Formula Weight
Melting Point (°C)
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing