Skip to Content
Merck
Heptenophos PESTANAL®, analytical standard

Heptenophos

Heptenophos PESTANAL®, analytical standard
Synonym(s):
7-Chlorobicyclo[3.2.0]hepta-2,6-dien-6-yl dimethyl phosphate
Empirical Formula (Hill Notation):
C9H12ClO4P
CAS Number:
Molecular Weight:
250.62
EC Number:

Products

Product Category
Brand
Formula Weight
Product Line
Purity
Format
Chemical Composition
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing