Skip to Content
Merck
vibrant-m

Haloperidol Related Compound B

vibrant-m
Synonym(s):
4-[4-(4-Chlorophenyl)-4-hydroxypiperidin-1-yl]-1-(2-fluorophenyl)butan-1-one
Empirical Formula (Hill Notation):
C21H23ClFNO2
CAS Number:
Molecular Weight:
375.86

Products

Shipping
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1