Skip to Content
Merck
γ-Valerolactone BioRenewable, anhydrous, DMF and NMP substitute, ≥99%

γ-Valerolactone

γ-Valerolactone BioRenewable, anhydrous, DMF and NMP substitute, ≥99%
Synonym(s):
γ-Valerolactone, γ-Methyl-γ-butyrolactone, 4,5-Dihydro-5-methyl-2(3H)-furanone, 4-Hydroxypentanoic acid lactone
Empirical Formula (Hill Notation):
C5H8O2
CAS Number:
Molecular Weight:
100.12

Products

Shipping
Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Color
Feature
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade