Skip to Content
Merck
Guanosine 5′-diphosphate sodium salt Type I, ≥96% (HPLC)

Guanosine 5′-diphosphate sodium salt

Guanosine 5′-diphosphate sodium salt Type I, ≥96% (HPLC)
Synonym(s):
GDP
Empirical Formula (Hill Notation):
C10H15N5O11P2
CAS Number:
Molecular Weight:
443.20

Products

Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing