Skip to Content
Merck
Guanidine thiocyanate for molecular biology, free-flowing, Redi-Dri™, ≥99.0%

Guanidine thiocyanate

Guanidine thiocyanate for molecular biology, free-flowing, Redi-Dri™, ≥99.0%
Synonym(s):
Guanidinium rhodanide, Guanidinium thiocyanate, Guanidine thiocyanate, Guanidine Thiocyanate
Linear Formula:
NH2C(=NH)NH2 · HSCN
CAS Number:
Molecular Weight:
118.16

Products

Shipping
Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Manufacturer
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Quality Segment
Application
Special Grade
Technique