Skip to Content
Merck
Growth Hormone Releasing Factor Fragment 1-29 amide human ≥97% (HPLC)

Growth Hormone Releasing Factor Fragment 1-29 amide human

Growth Hormone Releasing Factor Fragment 1-29 amide human ≥97% (HPLC)
Empirical Formula (Hill Notation):
C149H246N44O42S
CAS Number:
Molecular Weight:
3357.88

Products

Product Category
Brand
Formula Weight
Gene Alias
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing