![Glycolonitrile 55 wt. % in H2O](/deepweb/assets/sigmaaldrich/product/structures/202/639/c7531ea8-aa44-4cb2-bc1a-5969f147d9c5/640/c7531ea8-aa44-4cb2-bc1a-5969f147d9c5.png)
Glycolonitrile
![Glycolonitrile 55 wt. % in H2O](/deepweb/assets/sigmaaldrich/product/structures/202/639/c7531ea8-aa44-4cb2-bc1a-5969f147d9c5/640/c7531ea8-aa44-4cb2-bc1a-5969f147d9c5.png)
Synonym(s):
Glycolic acid nitrile solution, Formaldehyde cyanohydrin, Hydroxyacetonitrile
Linear Formula:
HOCH2CN
CAS Number:
Molecular Weight:
57.05
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing