
Glycolic acid ethoxylate oleyl ether

Synonym(s):
Oleth-10 carboxylic acid, Oleth-3 carboxylic acid, Oleth-6 carboxylic acid
Linear Formula:
CH3(CH2)xCH=CH(CH2)8(OCH2CH2)yOCH2CO2H, x = 5-7, y~2
CAS Number:
Molecular Weight:
901.47
Products
Shipping
Brand
Formula Weight
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing