Ginsenoside Rb1
Synonym(s):
2-O-β-Glucopyranosyl-(3β,12β)-20-[(6-O-β-D-glucopyranosyl-β-D-glucopyranosyl)oxy]-12-hydroxydammar-24-en-3-yl β-D-glucopyranoside, Arasaponin E1, Gynosaponin C, Gypenoside III, Notoginsenoside Rb1, Panaxoside Rb1, Sanchinoside E1, Sanchinoside Rb1
Empirical Formula (Hill Notation):
C54H92O23
CAS Number:
Molecular Weight:
1109.29
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3