![Ginkgolic acid C17:1 primary reference standard](/deepweb/assets/sigmaaldrich/product/structures/278/641/e08320d0-e279-4e26-b9a0-529b6cbbc29c/640/e08320d0-e279-4e26-b9a0-529b6cbbc29c.png)
Ginkgolic acid C17:1
![Ginkgolic acid C17:1 primary reference standard](/deepweb/assets/sigmaaldrich/product/structures/278/641/e08320d0-e279-4e26-b9a0-529b6cbbc29c/640/e08320d0-e279-4e26-b9a0-529b6cbbc29c.png)
Synonym(s):
6-[(10Z)-Heptadecenyl]salicylic acid, Ginkgolic acid II
Empirical Formula (Hill Notation):
C24H38O3
CAS Number:
Molecular Weight:
374.56
Products
Shipping
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3