Skip to Content
Merck
γ-Glu-Cys ≥80% (HPLC)

γ-Glu-Cys

γ-Glu-Cys ≥80% (HPLC)
Synonym(s):
γ-L-Glutamyl-L-cysteine, des-Gly-glutathione reduced form
Empirical Formula (Hill Notation):
C8H14N2O5S
CAS Number:
Molecular Weight:
250.27

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Chemical Composition
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing