Skip to Content
Merck
γ-Cyclodextrin powder, BioReagent, suitable for cell culture, ≥98%

γ-Cyclodextrin

γ-Cyclodextrin powder, BioReagent, suitable for cell culture, ≥98%
Synonym(s):
Cyclomaltooctaose, Cyclooctaamylose, Schardinger γ-Dextrin
Empirical Formula (Hill Notation):
C48H80O40
CAS Number:
Molecular Weight:
1297.12
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Technique