![gamma-Mangostin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/360/743/261e2b31-5447-4825-9d67-1a369347aaf8/640/261e2b31-5447-4825-9d67-1a369347aaf8.png)
gamma-Mangostin
![gamma-Mangostin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/360/743/261e2b31-5447-4825-9d67-1a369347aaf8/640/261e2b31-5447-4825-9d67-1a369347aaf8.png)
Synonym(s):
1,3,6,7-Tetrahydroxy-2,8-bis(3,3-dimethylallyl)xanthone, Demethylmangostin, Normangostin
Empirical Formula (Hill Notation):
C23H24O6
CAS Number:
Molecular Weight:
396.43
Products
Shipping
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing