
Fumaric acid-1,4-13C2

Linear Formula:
HOO13CCH=CH13COOH
Molecular Weight:
118.06
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing