Skip to Content
Merck
Fumaraldehyde bis(dimethyl acetal) 95%

Fumaraldehyde bis(dimethyl acetal)

Fumaraldehyde bis(dimethyl acetal) 95%
Synonym(s):
trans-1,1,4,4-Tetramethoxy-2-butene
Linear Formula:
(CH3O)2CHCH=CHCH(OCH3)2
CAS Number:
Molecular Weight:
176.21

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing