Skip to Content
Merck
(Formylmethyl)triphenylphosphonium chloride ≥97.0% (AT)

(Formylmethyl)triphenylphosphonium chloride

(Formylmethyl)triphenylphosphonium chloride ≥97.0% (AT)
Synonym(s):
2-Oxoethyltriphenylphosphonium chloride
Linear Formula:
HCOCH2P(Cl)(C6H5)3
CAS Number:
Molecular Weight:
340.78
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing