![Foramsulfuron PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/268/412/600188f2-dd2f-4a8d-b204-92a33ece06eb/640/600188f2-dd2f-4a8d-b204-92a33ece06eb.png)
Foramsulfuron
![Foramsulfuron PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/268/412/600188f2-dd2f-4a8d-b204-92a33ece06eb/640/600188f2-dd2f-4a8d-b204-92a33ece06eb.png)
Synonym(s):
2-[3-(4,6-Dimethoxy-2-pyrimidinyl)ureidosulfonyl]-4-(formamido)-N,N-dimethylbenzamide
Empirical Formula (Hill Notation):
C17H20N6O7S
CAS Number:
Molecular Weight:
452.44
Products
Shipping
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2