Skip to Content
Merck
Fluquinconazole PESTANAL®, analytical standard

Fluquinconazole

Fluquinconazole PESTANAL®, analytical standard
Synonym(s):
3-(2,4-Dichlorophenyl)-6-fluoro-2-(1H-1,2,4-trizol-1-yl)quinazolin-4(3H)-one, Castellan
Empirical Formula (Hill Notation):
C16H8Cl2FN5O
CAS Number:
Molecular Weight:
376.17
EC Number:

Products

Shipping
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing