Skip to Content
Merck
Flavone analytical standard

Flavone

Flavone analytical standard
Synonym(s):
2-Phenyl-4H-1-benzopyran-4-one, 2-Phenylchromone
Empirical Formula (Hill Notation):
C15H10O2
CAS Number:
Molecular Weight:
222.24

Products

Shipping
Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing