Skip to Content
Merck
Firocoxib VETRANAL®, analytical standard

Firocoxib

Firocoxib VETRANAL®, analytical standard
Synonym(s):
3-Cyclopropylmethoxy-4-(4-methanesulfonylphenyl)-5,5-dimethyl-5H-furan-2-one
Empirical Formula (Hill Notation):
C17H20O5S
CAS Number:
Molecular Weight:
336.40

Products

Shipping
Product Category
Brand
Color
Formula Weight
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing