![Ferrocene 98%](/deepweb/assets/sigmaaldrich/product/structures/298/230/88229b40-738a-4df7-a1bd-41948e99cd7b/640/88229b40-738a-4df7-a1bd-41948e99cd7b.png)
Ferrocene
![Ferrocene 98%](/deepweb/assets/sigmaaldrich/product/structures/298/230/88229b40-738a-4df7-a1bd-41948e99cd7b/640/88229b40-738a-4df7-a1bd-41948e99cd7b.png)
Synonym(s):
Bis(cyclopentadienyl)iron, Di(cyclopentadienyl)iron, Ferrocene
Linear Formula:
(C5H5)2Fe
CAS Number:
Molecular Weight:
186.03
Products
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Reagent Type
Core
Showing 1-2 of 2
Compare
Product Number
Description
Pricing