
Fenpiclonil

Empirical Formula (Hill Notation):
C11H6Cl2N2
CAS Number:
Molecular Weight:
237.08
Products
Shipping
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2