
Fenofibric acid

Synonym(s):
2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionic acid, Fenofibric acid
Empirical Formula (Hill Notation):
C17H15ClO4
Products
Brand
Formula Weight
Gene Alias
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Technique
Showing 1-1 of 1