Skip to Content
Merck
Fenaminosulf PESTANAL®, analytical standard

Fenaminosulf

Fenaminosulf PESTANAL®, analytical standard
Synonym(s):
4-Dimethylaminobenzenediazosulfonic acid sodium salt, p-Diazodimethylaniline sodium sulfonate, Sodium [4-(dimethylamino)phenyl]diazenesulfonate
Linear Formula:
C8H10N3O3SNa
CAS Number:
Molecular Weight:
251.24
EC Number:

Products

Shipping
Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing