Skip to Content
Merck
Ethyl myristate 99% (GC)

Ethyl myristate

Ethyl myristate 99% (GC)
Synonym(s):
Ethyl tetradecanoate, Myristic acid ethyl ester
Linear Formula:
CH3(CH2)12COOC2H5
CAS Number:
Molecular Weight:
256.42
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing