Skip to Content
Merck
Ethyl heptanoate natural, ≥98%, FG

Ethyl heptanoate

Ethyl heptanoate natural, ≥98%, FG
Synonym(s):
Ethyl enanthate
Linear Formula:
CH3(CH2)5COOC2H5
CAS Number:
Molecular Weight:
158.24
EC Number:

Products

Shipping
Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing