Skip to Content
Merck
Ethylenediaminetetraacetic acid tripotassium salt dihydrate puriss. p.a., ≥99.0% (KT)

Ethylenediaminetetraacetic acid tripotassium salt dihydrate

Ethylenediaminetetraacetic acid tripotassium salt dihydrate puriss. p.a., ≥99.0% (KT)
Synonym(s):
EDTA tripotassium salt, Edathamil, Tripotassium ethylenediaminetetraacetate dihydrate
Linear Formula:
(KOOCCH2)2NCH2CH2N(CH2COOK)CH2COOH · 2H2O
CAS Number:
Molecular Weight:
442.54

Products

Shipping
Product Category
Brand
Biological Source
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Reagent Type
Special Grade