Skip to Content
Merck
Ethyl decanoate ≥98%, FCC, FG

Ethyl decanoate

Ethyl decanoate ≥98%, FCC, FG
Synonym(s):
Capric acid ethyl ester, Ethyl caprate, Ethyl decanoate
Linear Formula:
CH3(CH2)8COOC2H5
CAS Number:
Molecular Weight:
200.32

Products

Shipping
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-4 of 4