![ethyl 2-N-butylacetoacetate AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/800/847/9241fe48-009f-44df-a2fe-60be8062e78e/640/9241fe48-009f-44df-a2fe-60be8062e78e.png)
ethyl 2-N-butylacetoacetate
![ethyl 2-N-butylacetoacetate AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/800/847/9241fe48-009f-44df-a2fe-60be8062e78e/640/9241fe48-009f-44df-a2fe-60be8062e78e.png)
Empirical Formula (Hill Notation):
C10H18O3
CAS Number:
Molecular Weight:
186.25
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing