
Ethyl 2-cyano-3,3-diphenylacrylate

Linear Formula:
(C6H5)2C=C(CN)CO2C2H5
CAS Number:
Molecular Weight:
277.32
Products
Shipping
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing