![Ethofumesate-2-keto PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/792/202/b4b5cd25-2985-45f3-a24f-3c6fdd781b12/640/b4b5cd25-2985-45f3-a24f-3c6fdd781b12.png)
Ethofumesate-2-keto
![Ethofumesate-2-keto PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/792/202/b4b5cd25-2985-45f3-a24f-3c6fdd781b12/640/b4b5cd25-2985-45f3-a24f-3c6fdd781b12.png)
Synonym(s):
3,3-Dimethyl-2-oxo-2,3-dihydrobenzofurane-5-yl methane sulfonate, Methane sulfonic acid 3,3-dimethyl-2-oxo-2,3-dihydrobenzofuran-5-yl ester
Empirical Formula (Hill Notation):
C11H12O5S
CAS Number:
Molecular Weight:
256.27
Products
Shipping
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing