Skip to Content
Merck
Ethanesulfonic acid sodium salt monohydrate ≥98.0% (T)

Ethanesulfonic acid sodium salt monohydrate

Ethanesulfonic acid sodium salt monohydrate ≥98.0% (T)
Synonym(s):
Sodium-ethanesulfonate monohydrate
Linear Formula:
CH3CH2SO3Na·H2O
CAS Number:
Molecular Weight:
150.13
EC Number:

Products

Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing