Skip to Content
Merck
Erucic acid analytical standard

Erucic acid

Erucic acid analytical standard
Synonym(s):
cis-13-Docosenoic acid, Prifac 2990
Linear Formula:
CH3(CH2)7CH=CH(CH2)11COOH
CAS Number:
Molecular Weight:
338.57
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Functional Group
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing