Skip to Content
Merck
Enrofloxacin European Pharmacopoeia (EP) Reference Standard

Enrofloxacin

Enrofloxacin European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Baytril, Enrofloxacin
Empirical Formula (Hill Notation):
C19H22FN3O3
CAS Number:
Molecular Weight:
359.39

Products

Shipping
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
Package Size
API Family
Reference Material Type
Special Grade
Technique