
ECO Brij® S10

Synonym(s):
Brij® S10, Polyethylene glycol octadecyl ether, Polyoxyethylene (10) stearyl ether
Linear Formula:
C18H37(OCH2CH2)nOH, n~10
CAS Number:
Products
Brand
Sustainability
Feature
Melting Point (°C)
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing