
DNOC

Synonym(s):
2-Methyl-4,6-dinitrophenol, 3,5-Dinitro-2-hydroxytoluene, 4,6-Dinitro-2-methylphenol, 4,6-Dinitro-o-cresol, DNOC
Empirical Formula (Hill Notation):
C7H6N2O5
CAS Number:
Molecular Weight:
198.13
Products
Shipping
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing