Skip to Content
Merck
DL-Valine ≥97%

DL-Valine

DL-Valine ≥97%
Synonym(s):
(±)-α-Aminoisovaleric acid, DL-2-Amino-3-methylbutanoic acid
Linear Formula:
(CH3)2CHCH(NH2)COOH
CAS Number:
Molecular Weight:
117.15
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Application
Markush Class
Markush Group
Optical Isomer
Organoleptic Class
Amino Acid
Reaction Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing