Skip to Content
Merck
DL-p-Hydroxyphenyllactic acid ≥97% (HPLC)

DL-p-Hydroxyphenyllactic acid

DL-p-Hydroxyphenyllactic acid ≥97% (HPLC)
Synonym(s):
3-(4-Hydroxyphenyl)-2-hydroxypropanoic acid
Empirical Formula (Hill Notation):
C9H10O4
CAS Number:
Molecular Weight:
182.17

Products

Shipping
Product Category
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing