Skip to Content
Merck
DL-Menthol analytical standard

(±)-Menthol

DL-Menthol analytical standard
Synonym(s):
Menthol, 2-Isopropyl-5-methylcyclohexanol, Hexahydrothymol
Empirical Formula (Hill Notation):
C10H20O
CAS Number:
Molecular Weight:
156.27
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing