Skip to Content
Merck
DL-Kynurenine certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

DL-Kynurenine

DL-Kynurenine certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
2-Amino-3-(2-aminobenzoyl)propionic acid, 2-Amino-4-(2-aminophenyl)-4-oxobutyric acid
Empirical Formula (Hill Notation):
C10H12N2O3
CAS Number:
Molecular Weight:
208.21

Products

Shipping
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade