DL-Glutamic acid monohydrate
Synonym(s):
(±)-2-Aminoglutaric acid, (±)-2-Aminopentanedioic acid
Linear Formula:
HOOCCH2CH2CH(NH2)COOH · H2O
Products
Shipping
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Amino Acid
Showing 1-1 of 1
Compare
Product Number
Description
Pricing