
DL-Dithiothreitol solution

Synonym(s):
(±)-threo-1,4-Dimercapto-2,3-butanediol solution, DTT
Empirical Formula (Hill Notation):
C4H10O2S2
CAS Number:
Molecular Weight:
154.25
Products
Shipping
Brand
Formula Weight
pH Value
Physical Form
Product Line
Research Area
Reagent Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing